Learn More
Cayman Chemical zngbr offIcnle 8Gngerol 10mg
SDP
Supplier:  Cayman Chemical 1184110 MG
Encompass_Preferred
A natural chemical that activates TRPV1 (EC50 = 5.0 µM), inhibits COX-2, and inhibits the growth of H. pylori in vitro; also augments wound healing, suppresses IL-2-induced proliferation of T lymphocytes, and potentiates β-agonist-induced relaxation of airway smooth muscle
Catalog No. 50-295-2126