Learn More
Apexbio Technology LLC Zofenopril calcium 81938-43-4 5mg
SDP
Supplier:  Apexbio Technology LLC B34685
Encompass_Preferred
Zofenopril calcium (CAS 81938-43-4) is a small-molecule inhibitor of angiotensin-converting enzyme (ACE) recognized for its antioxidative and cardioprotective actions It decreases oxidative stress by limiting the production of reactive oxygen species (ROS) and sustaining intracellular glutathione (GSH) levels thereby attenuating foam cell formation during early stages of atherosclerosis Experimental studies also indicate that Zofenopril calcium mitigates chronic myocardial injury induced by doxorubicin These properties make it a valuable agent for investigating oxidative stress atherogenesis and cardiotoxicity mechanisms in preclinical biomedical research
Catalog No. 50-000-94292