Learn More
Cayman Chemical ZofenoprIlcalcIum salt 10mg
SDP
Supplier:  Cayman Chemical 2952510 MG
Encompass_Preferred
A prodrug form of zofenoprilat; an ACE inhibitor (IC50 = 0.9 nM in heart tissue homogenates); reduces mean arterial blood pressure in 2K-1C rats and SHRs at 2.2, 6.6, and 22 mg/kg
Catalog No. 50-298-9134