Learn More
Medchemexpress LLC Zoledronic Acid | 118072-93-8 | 99.9% | 272.09 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1377750MG
Encompass_Preferred
Zoledronic Acid is a third-generation bisphosphonate with potent anti-resorptive activity. It inhibits osteoclast differentiation and apoptosis, and exhibits anti-cancer effects. It appears as a white to off-white solid.
  • Potent anti-resorptive activity
  • Inhibits osteoclast differentiation and apoptosis
  • Possesses anti-cancer effects
  • Ships at room temperature
  • Store at 4°C under nitrogen, or -80°C/-20°C in solvent under nitrogen
Catalog No. 50-003-51915