Learn More
Cayman Chemical ZolIflodacIn 10mg
SDP
Supplier:  Cayman Chemical 3420310 MG
Encompass_Preferred
A fluoroquinolone antibiotic; inhibits bacterial DNA gyrase supercoiling activity and topoisomerase IV DNA decatenation; active against 250 clinical isolates of N. gonorrhoeae, including XDR and MDR strains (MICs = 0.004-0.25 UG/ml); active against MRSA clinical isolates, as well as 10 clinical isolates EAch of C. trachomatis and C. pneumoniae (MIC50s = 0.125 and 0.5 UG/ml, respectively)
Catalog No. 50-295-0223