Learn More
Apexbio Technology LLC Zolmitriptan 139264-17-8 500mg
SDP
Supplier:  Apexbio Technology LLC B2261500
Encompass_Preferred
Zolmitriptan (CAS 139264-17-8) is a synthetic small molecule that functions as a selective agonist of serotonin 5-HT1B and 5-HT1D receptors By stimulating these receptors zolmitriptan modulates trigeminovascular neurotransmission leading to cranial vasoconstriction and inhibition of pro-inflammatory neuropeptide release These mechanisms underlie its ability to acutely alleviate migraine attacks both with and without aura as well as cluster headaches In biomedical research zolmitriptan serves as a tool compound for investigating serotonergic signaling pathways involved in nociception and vascular regulation associated with headache disorders
Catalog No. 50-000-94156