Learn More
Cayman Chemical ZolmItrIptn 10mg
SDP
Supplier:  Cayman Chemical 1587110 MG
Encompass_Preferred
A triptan that acts as an agonist of the serotonin (5-HT) receptors 5-HT1B and 5-HT1D (Kis = 2.2 and 0.92 nM, respectively); inhibits cAMP formation in blood vessels, resulting in vessel constriction
Catalog No. 50-297-2911