Learn More
Medchemexpress LLC Zoniporide hydrochloride hydrate | 863406-85-3 | MFCD04113213 | >98.0% | 374.82 g·mol⁻¹ | C17H19ClN6O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY105064D100MG
Encompass_Preferred
Zoniporide hydrochloride hydrate is a research-use small molecule that potently and selectively inhibits the sodium-hydrogen exchanger isoform 1 (NHE-1). It is used in cellular and pharmacological studies to probe NHE-1-mediated pathways such as cardioprotection and cell volume regulation. Available in solid form and as a DMSO solution for laboratory assays.
  • Potent NHE-1 inhibitor (human NHE-1 IC50 = 14 nM).
  • Shows >150-fold selectivity versus other NHE isoforms.
  • Inhibits ex vivo NHE-1-dependent platelet swelling (IC50 = 59 nM).
  • Supplied in solid and 10 mM DMSO solution formats for flexibility.
  • Intended for research use only.
Catalog No. 50-002-44273