Learn More
Apexbio Technology LLC Zonisamide 68291-97-4 200mg
SDP
Supplier:  Apexbio Technology LLC A5354200
Encompass_Preferred
Zonisamide (CAS 68291-97-4) is a small molecule antiepileptic agent characterized by its broad-spectrum anticonvulsant mechanisms It inhibits neuronal hyperexcitability primarily through blockade of voltage-gated sodium channels and T-type calcium channels Zonisamide also facilitates large-conductance calcium-activated potassium (BKCa) channel activity and modulates GABAergic glutamatergic and monoaminergic neurotransmission Due to its multifaceted pharmacological actions zonisamide is utilized in research focused on seizure disorders ion channel function and the modulation of central nervous system excitability
Catalog No. 50-000-97338