Learn More
Selleck Chemical LLC Zonisamide 25mg 68291-97-4 CI-912
SDP
Supplier:  Selleck Chemical LLC S144525MG
Encompass_Preferred
Zonisamide (CI-912) is a voltage-dependent sodium channel and T-type calcium channel blocker, used as an antiepileptic drug. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-768-2