Learn More
Andwin Scientific ZORBAX SBC18 3.0X5150MM
SDP
Supplier:  Andwin Scientific 863954302
Encompass_Preferred
ZORBAX SB-C18, Solvent Saver plus HPLC Column, Reversed-phase separations of acidic neutral basic compounds, 80angstrom Pore Size, 3.5um, 3.0 I.D. x 150mm L, pH 1 to 8, Carbon Load 10, 1/EA
Catalog No. NC9173710