Learn More
TARGETMOL CHEMICALS INC Zorifertinib 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3659200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Zorifertinib (AZD3759) is an orally active, effective and central nervous system-penetrant EGFR inhibitor. The IC50 values were 0.2, 0.3 and 0.2 nM for EGFR L858R mutant, TK wild-type, and Exon 19Del enzymes, respectively. Purity 99.36%
Catalog No. 50-288-0055