Learn More
Medchemexpress LLC Zr17-2 | 1263893-98-6 | 98.0% | 324.40 | C13H20N6O2S | 100MG
SDP
Supplier:  Medchemexpress LLC HY125972100MG
Encompass_Preferred
zr17-2 is a small-molecule modulator of the cannabinoid CB1 receptor reported to reduce oxidative stress-induced retinal cell death. It is supplied as a solid for biochemical and cellular research focused on cannabinoid signaling and neuroprotection studies.
  • Modulates cannabinoid CB1 receptor activity.
  • Reduces oxidative stress-induced retinal cell death in model systems.
  • High reported purity (98.0%).
  • Available in multiple research-scale pack sizes.
  • Supplied as a solid suitable for formulation in common solvents.
Catalog No. 50-002-36539