Learn More
Medchemexpress LLC HY-11013 5mg Medchemexpress, ZSET1446 CAS:887603-94-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY110135MG
Encompass_Preferred
Medchemexpress, HY-11013 5mg ZSET1446 CAS:887603-94-3 ZSET1446 is a novel cognitive enhancer that significantly improves learning deficits in various types of Alzheimer disease (AD) models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8753