Learn More
TARGETMOL CHEMICALS INC ZT-12-037-01 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T559710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zt-12-037-01 is a ATP competitive STK19 inhibitor(IC50 values of 23.96 nM and 27.94 nM for STK19 (WT) and STK19 (D89N)respectively). purity: 98%
Catalog No. 50-257-6670