Learn More
Selleck Chemical LLC ZT-12-037-01 S8864-5MG
SDP
Supplier:  Selleck Chemical LLC S88645MG
Encompass_Preferred
ZT-12-037-01 is a specific and ATP-competitive STK19 inhibitor with IC50s of 23 96 nM and 27 94 nM for STK19 (WT) and STK19 (D89N) respectively It displays extremely high kinase selectivity using KINOMEscan against a panel of 468 diverse kinases using an in vitro ATP-site competition binding assay at 1 M
Catalog No. 50-000-25034