Learn More
Selleck Chemical LLC ZT-1a E1948-100MG
SDP
Supplier:  Selleck Chemical LLC E1948100MG
Encompass_Preferred
ZT-1a is a potent and selective non-ATP-competitive inhibitor of STE20/SPS1-related proline-alanine-rich kinase (SPAK) with an IC50 of 44 3 M It can exhibit therapeutic potential for brain disorders linked to impaired ionic homeostasis in research
Catalog No. 50-000-39352