Learn More
Medchemexpress LLC ZT 52656A hydrochloride | 115730-24-0 | 100.0% | 390.87 g/mol | C19H26ClF3N2O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1015821MG
Encompass_Preferred
ZT 52656A hydrochloride is the hydrochloride salt of a selective kappa opioid receptor agonist supplied for laboratory research. It is used to investigate kappa-mediated analgesia, including prevention or alleviation of ocular pain, and is intended for preclinical in vitro and in vivo studies. Not for human or clinical use.
  • Selective kappa opioid receptor agonist.
  • Used in ocular pain models to assess analgesic effects.
  • High purity (99.98%) suitable for research applications.
  • Solid, white to off-white physical form.
  • Hydrochloride salt for improved stability and solubility.
  • Available in small research pack sizes for preclinical work.
Catalog No. 50-002-48059