Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zt 52656a hydrochloride | 115730-24-0 | MFCD00673899 | 100.0% | 390.87 g·mol⁻¹ | C19H26ClF3N2O | 100 MG
SDP

Supplier:  Medchemexpress LLC HY101582100MG

Encompass_Preferred

ZT 52656A hydrochloride is the hydrochloride salt of a selective kappa opioid receptor agonist used in research to investigate kappa-mediated analgesia and the prevention or alleviation of ocular pain. Supplied as a white to off-white solid for laboratory applications, it is intended for research use only and not for clinical or human use.

  • Selective kappa opioid receptor agonist.
  • High reported purity (99.98%).
  • Molecular weight 390.87 g·mol⁻¹.
  • White to off-white solid suitable for analytical and in vivo studies.
  • Recommended storage: 4°C sealed; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Available in multiple package sizes, including 100 mg.

Catalog No. 50-002-47901


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.