Learn More
Medchemexpress LLC Zuclopenthixol-d4 succinate | 1246833-97-5 | 99.0% | C26H27D4ClN2O5S | 1 MG
SDP
Supplier:  Medchemexpress LLC HYA0163S1MG
Encompass_Preferred
Zuclopenthixol-d4 succinate is the deuterium labeled Zuclopenthixol. Zuclopenthixol is a thioxanthene derivative which acts as a mixed dopamine D1/D2 receptor antagonist. It is for research use only and not sold to patients. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32406