Learn More
Medchemexpress LLC HY-N7394A 1mg Medchemexpress, (-)-Zuonin A CAS:84709-25-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7394A1MG
Encompass_Preferred
Medchemexpress, HY-N7394A 1mg (-)-Zuonin A CAS:84709-25-1 (-)-Zuonin A (D-Epigalbacin), a naturally occurring lignin, is a potent, selective JNKs inhibitor, with IC50s of 1.7 μM, 2.9 μM and 1.74 μM for JNK1, JNK2 and JNK3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2994