Learn More
Zymo Research Corporation Zymo-Spin™ IC-S (50 Pack)
SDP
Supplier:  Zymo Research Corporation C101550
Encompass_Preferred
The Zymo-Spin IC-S features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g of DNA, and 10 g of RNA, in 6 l eluate. Capacity is 800 l.
Catalog No. 50-203-6997