Learn More
Zymo Research Corporation ZymoBIOMICS 96 MagBead DNA Kit – Dx (Lysis Tubes) (2 x 96 preps)
SDP
Supplier:  Zymo Research Corporation D4308E
Encompass_Preferred
The ZymoBIOMICS DNA Kits are designed for purifying DNA from a variety of sample inputs that is immediately ready for microbiome or metagenome analyses. The ZymoBIOMICS lysis system eliminates bias associated with unequal lysis efficiencies of different organisms (e.g. Gram-negative/positive bacteria, fungus, protozoans, and algae), making it ideal for microbial community profiling. Uniform mechanical lysis of all microbes is achieved by bead beating with the innovative ultra-high density BashingBeads. This kit is equipped with our OneStep PCR Inhibitor removal technology, enabling PCR amplification from DNA derived from inhibitor-rich environmental samples. Purified DNA is ideal for all downstream applications including PCR, arrays, 16S rRNA gene sequencing, and shotgun sequencing. DNA Size is 15-20 kb.
Catalog No. NC2233801