Assay Details
| Gene Symbol: SIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128589.2 | NP_180595.1 | 1 | 1561 | 109 | |
| GenBank mRNA | AF339146.1 | - | 1 | 1299 | 109 | |
| BT002478.1 | - | 1 | 1561 | 109 | ||
More Information
Set Membership: |
|

SIP4
Genomic Map