Assay Details
| Gene Symbol: ISU3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116646.3 | NP_192317.1 | 2 - 3 | 443 | 61 | |
| GenBank mRNA | AJ866967.1 | - | 2 - 3 | 376 | 61 | |
| BT030662.1 | - | 2 - 3 | 348 | 61 | ||
| DQ056643.1 | - | 2 - 3 | 348 | 61 | ||
More Information
Set Membership: |
|

ISU3
Genomic Map