Assay Details
| Gene Symbol: CIPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036350.1 | NP_001031427.1 | 7 - 8 | 801 | 106 | |
| NM_001336096.1 | NP_001318295.1 | 7 - 8 | 807 | 106 | ||
| NM_179761.2 | NP_850092.1 | 7 - 8 | 801 | 106 | ||
| NM_179763.4 | NP_850094.1 | 8 - 9 | 906 | 106 | ||
| NM_179764.3 | NP_850095.2 | 8 - 9 | 979 | 106 | ||
| GenBank mRNA | AF367290.1 | - | 7 - 8 | 801 | 106 | |
| AY059163.1 | - | 7 - 8 | 683 | 106 | ||
| AY091098.1 | - | 7 - 8 | 794 | 106 | ||
| AY142671.1 | - | 7 - 8 | 683 | 106 | ||
| AY266298.1 | - | 7 - 8 | 683 | 106 | ||
More Information
Set Membership: |
|

CIPK3
Genomic Map