Assay Details
| Gene Symbol: AT3G19390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112826.4 | NP_566633.1 | 3 - 4 | 1031 | 74 | |
| GenBank mRNA | AY062725.1 | - | 3 - 4 | 867 | 74 | |
| AY093350.1 | - | 3 - 4 | 832 | 74 | ||
More Information
Set Membership: |
|

AT3G19390
Genomic Map