Assay Details
| Gene Symbol: CIPK21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345268.1 | NP_001330674.1 | 1 - 2 | 506 | 86 | |
| NM_125144.4 | NP_568860.1 | 1 - 2 | 580 | 86 | ||
| GenBank mRNA | AY034100.1 | - | 1 - 2 | 234 | 86 | |
More Information
Set Membership: |
|

CIPK21
Genomic Map