Assay Details
| Gene Symbol: UBP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345687.1 | NP_001332735.1 | 1 - 2 | 556 | 75 | |
| NM_125943.2 | NP_201348.1 | 1 - 2 | 228 | 75 | ||
More Information
Set Membership: |
|

UBP17
Genomic Map