Assay Details
| Gene Symbol: CLPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124714.4 | NP_568792.1 | 3 - 4 | 999 | 63 | |
| GenBank mRNA | AY035103.1 | - | 3 - 4 | 850 | 63 | |
| AY142561.1 | - | 3 - 4 | 706 | 63 | ||
More Information
Set Membership: |
|

CLPX
Genomic Map