Assay Details
| Gene Symbol: AT2G42940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129855.3 | NP_181822.1 | 1 | 1066 | 153 | |
| GenBank mRNA | BT010721.1 | - | 1 | 1023 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G42940| Gene Symbol: AT2G42940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129855.3 | NP_181822.1 | 1 | 1066 | 153 | |
| GenBank mRNA | BT010721.1 | - | 1 | 1023 | 153 | |
Set Membership: |
|