Assay Details
| Gene Symbol: NDUFA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176655.2 | NP_788828.1 | 5 - 6 | 667 | 57 | |
| GenBank mRNA | BC102913.1 | - | 4 - 5 | 596 | 57 | |
| X59419.1 | - | 667 | 57 | |||
More Information
Set Membership: |
|

NDUFA10
Genomic Map