Assay Details
| Gene Symbol: MRPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014932.1 | NP_001014932.1 | 4 - 5 | 559 | 71 | |
| GenBank mRNA | BC110214.1 | - | 4 - 5 | 570 | 71 | |
| BT021538.1 | - | 4 - 5 | 559 | 71 | ||
More Information
Set Membership: |
|

MRPL11
Genomic Map