Assay Details
| Gene Symbol: PPP1R7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034238.2 | NP_001029410.1 | 1 - 2 | 70 | 61 | |
| GenBank mRNA | BC102228.1 | - | 1 - 2 | 72 | 61 | |
More Information
Set Membership: |
|

PPP1R7
Genomic Map