Assay Details
| Gene Symbol: POLR2K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037623.3 | NP_001032712.1 | 2 - 3 | 145 | 147 | |
| NM_001303535.1 | NP_001290464.1 | 2 - 3 | 178 | 147 | ||
| NM_001303536.1 | NP_001290465.1 | 2 - 3 | 307 | 147 | ||
| GenBank mRNA | BC103450.1 | - | 2 - 3 | 119 | 147 | |
| DV801064.1 | - | 2 - 3 | 158 | 147 | ||
| DY069276.1 | - | 2 - 3 | 254 | 147 | ||
| EE222335.1 | - | 2 - 3 | 146 | 147 | ||
| EV627502.1 | - | 2 - 3 | 163 | 147 | ||
More Information
Set Membership: |
|

POLR2K
Genomic Map