Assay Details
| Gene Symbol: POLR2K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037623.3 | NP_001032712.1 | 3 - 4 | 226 | 87 | |
| NM_001303535.1 | NP_001290464.1 | 3 - 4 | 259 | 87 | ||
| NM_001303536.1 | NP_001290465.1 | 3 - 4 | 388 | 87 | ||
| GenBank mRNA | BC103450.1 | - | 3 - 4 | 200 | 87 | |
| DV801064.1 | - | 3 - 4 | 239 | 87 | ||
| DY069276.1 | - | 3 - 4 | 335 | 87 | ||
| EE222335.1 | - | 3 - 4 | 227 | 87 | ||
| EV627502.1 | - | 3 - 4 | 244 | 87 | ||
More Information
Set Membership: |
|

POLR2K
Genomic Map