Assay Details
| Gene Symbol: POLR3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076962.2 | NP_001070430.1 | 5 - 6 | 895 | 58 | |
| GenBank mRNA | BC111227.1 | - | 5 - 6 | 896 | 58 | |
More Information
Set Membership: |
|

POLR3H
Genomic Map