Assay Details
| Gene Symbol: POLR3GL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075213.1 | NP_001068681.1 | 4 - 5 | 442 | 77 | |
| GenBank mRNA | BC114765.1 | - | 4 - 5 | 442 | 77 | |
More Information
Set Membership: |
|

POLR3GL
Genomic Map