Assay Details
| Gene Symbol: ISCU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075683.2 | NP_001069151.1 | 1 - 2 | 169 | 120 | |
| GenBank mRNA | BC118409.1 | - | 1 - 2 | 170 | 120 | |
More Information
Set Membership: |
|

ISCU
Genomic Map