Assay Details
| Gene Symbol: RHOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078124.2 | NP_001071592.1 | 1 - 2 | 317 | 83 | |
| GenBank mRNA | BC105439.1 | - | 1 - 2 | 318 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105439.1, NM_001078124.2 transcripts.
Set Membership: |
|

RHOH
Genomic Map