Assay Details
| Gene Symbol: PRSS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076931.1 | NP_001070399.1 | 5 - 6 | 792 | 95 | |
| GenBank mRNA | BC123568.1 | - | 5 - 6 | 792 | 95 | |
More Information
Set Membership: |
|

PRSS8
Genomic Map