Assay Details
| Gene Symbol: UBR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082601.1 | NP_001076070.1 | 1 - 2 | 176 | 63 | |
| GenBank mRNA | BC133370.1 | - | 1 - 2 | 176 | 63 | |
More Information
Set Membership: |
|

UBR7
Genomic Map