Assay Details
| Gene Symbol: NDOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046158.2 | NP_001039623.2 | 2 - 3 | 307 | 74 | |
| GenBank mRNA | BC112851.1 | - | 2 - 3 | 307 | 74 | |
| BT025363.1 | - | 2 - 3 | 235 | 74 | ||
| DV838559.1 | - | 2 - 3 | 307 | 74 | ||
More Information
Set Membership: |
|

NDOR1
Genomic Map