Assay Details
| Gene Symbol: rpl-32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267265.4 | NP_001254194.1 | 2 - 3 | 257 | 73 | |
| NM_001267266.3 | NP_001254195.1 | 1 - 2 | 93 | 73 | ||
More Information
Set Membership: |
|

rpl-32
Genomic Map