Assay Details
| Gene Symbol: rpb-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065646.8 | NP_498047.2 | 7 - 8 | 3015 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpb-2| Gene Symbol: rpb-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065646.8 | NP_498047.2 | 7 - 8 | 3015 | 78 | |
Set Membership: |
|