Assay Details
| Gene Symbol: mca-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028255.4 | NP_001023426.1 | 5 - 6 | 784 | 112 | |
| NM_001028256.7 | NP_001023427.1 | 5 - 6 | 873 | 112 | ||
| NM_001268480.3 | NP_001255409.1 | 2 - 3 | 294 | 112 | ||
| NM_069308.6 | NP_501709.2 | 5 - 6 | 874 | 112 | ||
| GenBank mRNA | AJ223616.1 | - | 5 - 6 | 806 | 112 | |
| AY438644.1 | - | 5 - 6 | 874 | 112 | ||
| AY438645.1 | - | 5 - 6 | 846 | 112 | ||
More Information
Set Membership: |
|

mca-1
Genomic Map