Assay Details
| Gene Symbol: mca-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028255.4 | NP_001023426.1 | 8 - 9 | 3554 | 71 | |
| NM_001028256.7 | NP_001023427.1 | 8 - 9 | 3643 | 71 | ||
| NM_001268480.3 | NP_001255409.1 | 5 - 6 | 3064 | 71 | ||
| NM_069308.6 | NP_501709.2 | 8 - 9 | 3644 | 71 | ||
| GenBank mRNA | AJ223616.1 | - | 8 - 9 | 3576 | 71 | |
| AY438644.1 | - | 8 - 9 | 3644 | 71 | ||
| AY438645.1 | - | 8 - 9 | 3616 | 71 | ||
More Information
Set Membership: |
|

mca-1
Genomic Map