Assay Details
| Gene Symbol: try-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073020.5 | NP_505421.3 | 5 - 6 | 838 | 136 | |
| GenBank mRNA | JN651275.1 | - | 5 - 6 | 838 | 136 | |
| JN651276.1 | - | 5 - 6 | 869 | 136 | ||
| JN651277.1 | - | 4 - 5 | 747 | 136 | ||
More Information
Set Membership: |
|

try-5
Genomic Map