Assay Details
| Gene Symbol: lgc-40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076027.7 | NP_508428.2 | 7 - 8 | 1140 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lgc-40| Gene Symbol: lgc-40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076027.7 | NP_508428.2 | 7 - 8 | 1140 | 101 | |
Set Membership: |
|