Assay Details
| Gene Symbol: SOCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031631.1 | NP_001026801.1 | 463 | 100 | ||
| GenBank mRNA | AB164434.1 | - | 463 | 100 | ||
| AY485997.1 | - | 415 | 100 | |||
More Information
Set Membership: |
|

SOCS3
Genomic Map